Tetrakis(triphenylphosphin)platin(0) 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Tetrakis(triphenylphosphine)platinum
223960010
223960050
ACRO223960010EA
127
EUR
ACRO223960010
ACRO223960050
Tetrakis(triphenylphosphin)platin(0) 98%
Tetrakis(triphenylphosphin)platin(0)
Formel:
C₇₂H₆₀P₄Pt Schmelzpunkt: 159…160 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00010014 CAS-Nummer: 14221-02-4 EINECS: 238-087-4 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Carbon (C) | 67.8 to 71.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...