Tetrafluorborsäure 50% (w/w) in Wasser
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
196700250
196705000
196700025
ACRO196700250EA
33.5
EUR
ACRO196700250
ACRO196705000
ACRO196700025
Tetrafluorborsäure 50% (w/w) in Wasser
Formel:
HBF₄ Schmelzpunkt: –90 °C Dichte: 1,41 g/cm³ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011345 CAS-Nummer: 16872-11-0 EINECS: 240-898-3 UN-Nummer: 1775 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Solution (May develop some turbidity or precipitate) |
Titration with NaOH | 48.0 to 51.0 % |
Chloride (Cl) | ≤50 ppm |
Sulfate (SO4) | ≤200 ppm |
Iron (Fe) | ≤50 ppm |
Heavy metals (as Pb) | ≤500 ppm |
Free acid | ≤2.5 % (free boric acid) |
Fluorosilicic acid | ≤200 ppm (H2SiF6) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...