Natrium-L(+)-glutamat Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

49621-1KG 49621-250G
SIAL49621-1KGEA 72 EUR
SIAL49621-1KG SIAL49621-250G
Natrium-L(+)-glutamat Monohydrat, Sigma-Aldrich®
Natrium-L(+)-glutamat Monohydrat
Formel: C₅H₈NNaO₄·H₂O
Molekulargewicht: 187.13 g/mol
Schmelzpunkt: 225…240 °C
Flammpunkt: >225 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150138
CAS-Nummer: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR