Natrium-L(+)-glutamat Monohydrat, EMPROVE® ESSENTIAL FCC, E621, NF, reinst suitable for use as excipient, SAFC®
Lieferant: MERCK PRODUCTION CHEMICALS
Synonyme: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate
1.06445.9029 1.06445.1000
1.06445.9029EA 0 EUR
1.06445.9029 1.06445.1000
Natrium-L(+)-glutamat Monohydrat, EMPROVE® ESSENTIAL FCC, E621, NF, reinst suitable for use as excipient, SAFC®
Natrium-L(+)-glutamat Monohydrat
Formel: C₅H₈NNaO₄·H₂O
Molekulargewicht: 187.13 g/mol
Schmelzpunkt: 225…240 °C
Flammpunkt: >225 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150138
CAS-Nummer: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278