5-Fluoro-4-iodopyridin-2-amine 98%

Lieferant: Apollo Scientific
Achtung

PC912273-1G PC912273-5G PC912273-10G
APOSPC912273-1GEA 84.1 EUR
APOSPC912273-1G APOSPC912273-5G APOSPC912273-10G
5-Fluoro-4-iodopyridin-2-amine 98%
5-Fluoro-4-iodopyridin-2-amine
Formel: C₅H₄FIN₂
MDL Number: MFCD28722380
CAS-Nummer: 1649470-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR