4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-fluoro-5(trifluoromethyl)phenylboronic acid, 4-Bromo-2-fluoro-5-(trifluoromethyl)benzeneboronic acid
PC900987-250MG
PC900987-1G
PC900987-5G
APOSPC900987-250MGEA
84.1
EUR
APOSPC900987-250MG
APOSPC900987-1G
APOSPC900987-5G
4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid
Formel:
C₇H₄BBrF₄O₂ |
MDL Number:
MFCD20441807 CAS-Nummer: 1451393-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...