2-Bromo-5-iodopyridin-4-amine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315895-250MG
OR315895-1G
APOSOR315895-250MGEA
288
EUR
APOSOR315895-250MG
APOSOR315895-1G
2-Bromo-5-iodopyridin-4-amine 95%
2-Bromo-5-iodopyridin-4-amine
Formel:
C₅H₄BrIN₂ |
MDL Number:
MFCD27959281 CAS-Nummer: 1300750-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...