5-Bromo-2-(cyclopropyl)thiophene-3-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR315887-250MG
OR315887-1G
APOSOR315887-250MGEA
219
EUR
APOSOR315887-250MG
APOSOR315887-1G
5-Bromo-2-(cyclopropyl)thiophene-3-carboxylic acid 95%
5-Bromo-2-(cyclopropyl)thiophene-3-carboxylic acid
Formel:
C₈H₇BrO₂S Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD32689355 CAS-Nummer: 2594436-03-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...