4-Acetyl-3-bromothiophene 95%

Lieferant: Apollo Scientific
Achtung

OR315854-250MG OR315854-1G
APOSOR315854-250MGEA 284 EUR
APOSOR315854-250MG APOSOR315854-1G
4-Acetyl-3-bromothiophene 95%
4-Acetyl-3-bromothiophene
Formel: C₆H₅BrOS
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD18451709
CAS-Nummer: 35717-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR