3-(Cyclopropyl)-2-nitroaniline 95%

Lieferant: Apollo Scientific
Achtung

OR315838-250MG OR315838-1G OR315838-5G
APOSOR315838-250MGEA 190 EUR
APOSOR315838-250MG APOSOR315838-1G APOSOR315838-5G
3-(Cyclopropyl)-2-nitroaniline 95%
3-(Cyclopropyl)-2-nitroaniline
Formel: C₉H₁₀N₂O₂
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD32669824
CAS-Nummer: 2097553-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR