4-Bromo-2-(difluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC57772-250MG PC57772-1G PC57772-5G
APOSPC57772-250MGEA 190 EUR
APOSPC57772-250MG APOSPC57772-1G APOSPC57772-5G
4-Bromo-2-(difluoromethyl)benzyl bromide 95%
4-Bromo-2-(difluoromethyl)benzyl bromide
Formel: C₈H₆Br₂F₂
MDL Number: MFCD18391074
CAS-Nummer: 1261448-03-6
UN-Nummer: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR