3-Chloro-5-fluoro-2-methoxybenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC57531-1G
PC57531-5G
APOSPC57531-1GEA
522
EUR
APOSPC57531-1G
APOSPC57531-5G
3-Chloro-5-fluoro-2-methoxybenzoic acid
3-Chloro-5-fluoro-2-methoxybenzoic acid
Formel:
C₈H₆ClFO₃ Molekulargewicht: 204.583 g/mol |
MDL Number:
MFCD22415153 CAS-Nummer: 1554882-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...