2-Fluorpyridin 97%

Lieferant: Apollo Scientific
Gefahr

PC57491-50G PC57491-100G PC57491-250G
APOSPC57491-50GEA 26.1 EUR
APOSPC57491-50G APOSPC57491-100G APOSPC57491-250G
2-Fluorpyridin 97%

JOC., 53, 2740 (1998): A useful precursor to a large range of 2,3-disubstituted pyridines. Fluorine is activated to nucleophilic displacement & C-3 metallation is possible.

Formel: C₅H₄FN
Siedepunkt: 124 - 126 °C
Dichte: 1,128
Flammpunkt: 24 °C
MDL Number: MFCD00006224
CAS-Nummer: 372-48-5
UN-Nummer: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR