4'-Bromo-2'-(difluoromethyl)acetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[4-Bromo-2-(difluoromethyl)phenyl]ethanone
PC57444-250MG
PC57444-1G
PC57444-5G
APOSPC57444-250MGEA
203
EUR
APOSPC57444-250MG
APOSPC57444-1G
APOSPC57444-5G
4'-Bromo-2'-(difluoromethyl)acetophenone 95%
4'-Bromo-2'-(difluoromethyl)acetophenone
Formel:
C₉H₇BrF₂O |
CAS-Nummer:
2386658-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...