4-Bromo-2-fluoro-3-(trifluoromethyl)phenol 95%

Lieferant: Apollo Scientific
Gefahr

PC57421-250MG PC57421-1G PC57421-5G
APOSPC57421-250MGEA 174 EUR
APOSPC57421-250MG APOSPC57421-1G APOSPC57421-5G
4-Bromo-2-fluoro-3-(trifluoromethyl)phenol 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)phenol
Formel: C₇H₃BrF₄O
MDL Number: MFCD28739074
CAS-Nummer: 1807145-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR