5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC57130-250MG
PC57130-1G
PC57130-5G
APOSPC57130-250MGEA
125
EUR
APOSPC57130-250MG
APOSPC57130-1G
APOSPC57130-5G
5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene 95%
5-Bromo-1-fluoro-2-iodo-3-(trifluoromethyl)benzene
Formel:
C₇H₂BrF₄I |
MDL Number:
MFCD33404151 CAS-Nummer: 2385351-70-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...