2-Bromo-4-chloro-5-fluorobenzonitrile 95%

Lieferant: Apollo Scientific
Gefahr

PC57125-250MG PC57125-1G PC57125-5G
APOSPC57125-250MGEA 144 EUR
APOSPC57125-250MG APOSPC57125-1G APOSPC57125-5G
2-Bromo-4-chloro-5-fluorobenzonitrile 95%
2-Bromo-4-chloro-5-fluorobenzonitrile
Formel: C₇H₂BrClFN
MDL Number: MFCD30184437
CAS-Nummer: 1893432-29-5
UN-Nummer: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR