6-Bromo-2-fluoro-3-iodobenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

PC56996-250MG PC56996-1G PC56996-5G
APOSPC56996-250MGEA 174 EUR
APOSPC56996-250MG APOSPC56996-1G APOSPC56996-5G
6-Bromo-2-fluoro-3-iodobenzotrifluoride 95%
6-Bromo-2-fluoro-3-iodobenzotrifluoride
Formel: C₇H₂BrF₄I
CAS-Nummer: 2383867-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR