Methyl 5-chloro-4-fluoro-2-(trifluoromethyl)benzoate 95%

Lieferant: Apollo Scientific
Achtung

PC56992-250MG PC56992-1G PC56992-5G
APOSPC56992-250MGEA 192 EUR
APOSPC56992-250MG APOSPC56992-1G APOSPC56992-5G
Methyl 5-chloro-4-fluoro-2-(trifluoromethyl)benzoate 95%
Methyl 5-chloro-4-fluoro-2-(trifluoromethyl)benzoate
Formel: C₉H₅ClF₄O₂
MDL Number: MFCD28752648
CAS-Nummer: 1805524-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR