2-Bromo-4-chloro-5-fluorobenzotrifluoride 95%

Lieferant: Apollo Scientific
Achtung

PC56991-250MG PC56991-1G PC56991-5G
APOSPC56991-250MGEA 125 EUR
APOSPC56991-250MG APOSPC56991-1G APOSPC56991-5G
2-Bromo-4-chloro-5-fluorobenzotrifluoride 95%
2-Bromo-4-chloro-5-fluorobenzotrifluoride
Formel: C₇H₂BrClF₄
CAS-Nummer: 2166961-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR