5-(Difluoromethyl)nicotinic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Difluoromethyl)pyridine-3-carboxylic acid
PC55851-250MG
PC55851-1G
APOSPC55851-250MGEA
395
EUR
APOSPC55851-250MG
APOSPC55851-1G
5-(Difluoromethyl)nicotinic acid 95%
5-(Difluoromethyl)nicotinic acid
Formel:
C₇H₅F₂NO₂ |
MDL Number:
MFCD18256211 CAS-Nummer: 1256817-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...