2'-Bromo-4'-(trifluoromethoxy)-2,2,2-trifluoroacetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-Bromo-4-(trifluoromethoxy)phenyl]-2,2,2-trifluoroethanone
PC53635-250MG
PC53635-1G
PC53635-5G
APOSPC53635-250MGEA
190
EUR
APOSPC53635-250MG
APOSPC53635-1G
APOSPC53635-5G
2'-Bromo-4'-(trifluoromethoxy)-2,2,2-trifluoroacetophenone 95%
2'-Bromo-4'-(trifluoromethoxy)-2,2,2-trifluoroacetophenone
Formel:
C₉H₃BrF₆O₂ |
MDL Number:
MFCD34169323 CAS-Nummer: 2676196-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...