Methyl 3-(1,1-difluoroethyl)benzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53632-250MG
PC53632-1G
PC53632-5G
APOSPC53632-250MGEA
174
EUR
APOSPC53632-250MG
APOSPC53632-1G
APOSPC53632-5G
Methyl 3-(1,1-difluoroethyl)benzoate 95%
Methyl 3-(1,1-difluoroethyl)benzoate
Formel:
C₁₀H₁₀F₂O₂ |
MDL Number:
MFCD30717684 CAS-Nummer: 878745-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...