4-Bromo-5-fluoro-2-iodobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53388-250MG
PC53388-1G
PC53388-5G
APOSPC53388-250MGEA
154
EUR
APOSPC53388-250MG
APOSPC53388-1G
APOSPC53388-5G
4-Bromo-5-fluoro-2-iodobenzaldehyde 95%
4-Bromo-5-fluoro-2-iodobenzaldehyde
Formel:
C₇H₃BrFIO |
MDL Number:
MFCD18910206 CAS-Nummer: 1379347-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...