4-Bromo-5-fluoro-2-iodobenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC53386-250MG
PC53386-1G
PC53386-5G
APOSPC53386-250MGEA
154
EUR
APOSPC53386-250MG
APOSPC53386-1G
APOSPC53386-5G
4-Bromo-5-fluoro-2-iodobenzoic acid 95%
4-Bromo-5-fluoro-2-iodobenzoic acid
Formel:
C₇H₃BrFIO₂ |
MDL Number:
MFCD30297369 CAS-Nummer: 1870155-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...