2,3-Difluoro-6-(trifluoromethoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC502488-250MG PC502488-1G PC502488-5G
APOSPC502488-250MGEA 174 EUR
APOSPC502488-250MG APOSPC502488-1G APOSPC502488-5G
2,3-Difluoro-6-(trifluoromethoxy)benzoic acid 95%
2,3-Difluoro-6-(trifluoromethoxy)benzoic acid
Formel: C₈H₃F₅O₃
MDL Number: MFCD31561163
CAS-Nummer: 2244076-73-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR