2,6-Difluoro-3-(trifluoromethoxy)benzyl alcohol 95%

Lieferant: Apollo Scientific
Achtung

PC502455-250MG PC502455-1G PC502455-5G
APOSPC502455-250MGEA 174 EUR
APOSPC502455-250MG APOSPC502455-1G APOSPC502455-5G
2,6-Difluoro-3-(trifluoromethoxy)benzyl alcohol 95%
2,6-Difluoro-3-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₅F₅O₂
MDL Number: MFCD28399110
CAS-Nummer: 1800231-21-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR