Pentafluoroethane sulphonic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Perfluoroethane sulphonate, 1,1,2,2,2-Pentafluoroethanesulphonic acid
PC49706-250MG
PC49706-1G
APOSPC49706-250MGEA
331
EUR
APOSPC49706-250MG
APOSPC49706-1G
Pentafluoroethane sulphonic acid 97%
Pentafluoroethane sulphonic acid
May be present as the hydrate.
Formel:
C₂HF₅O₃S Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD01861156 CAS-Nummer: 354-88-1 UN-Nummer: 2586 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...