Calcium hexafluorophosphate

Lieferant: Apollo Scientific
Gefahr

PC53079-1G PC53079-25G PC53079-5G
APOSPC53079-1GEA 79.9 EUR
APOSPC53079-1G APOSPC53079-25G APOSPC53079-5G
Calcium hexafluorophosphate
Calcium hexafluorophosphate
Formel: Ca₂PF₆
MDL Number: MFCD28963758
CAS-Nummer: 78415-39-1
UN-Nummer: 3260
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR