4-Bromo-6-fluoro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC49201-1G PC49201-250MG
APOSPC49201-1GEA 290 EUR
APOSPC49201-1G APOSPC49201-250MG
4-Bromo-6-fluoro-1H-benzimidazole
4-Bromo-6-fluoro-1H-benzimidazole
Formel: C₇H₄BrFN₂
Schmelzpunkt: 141…143 °C
MDL Number: MFCD15071663
CAS-Nummer: 1245642-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR