2-(Difluoromethyl)-1-fluoro-4-nitrobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC912795-1G
PC912795-5G
PC912795-25G
APOSPC912795-1GEA
53.8
EUR
APOSPC912795-1G
APOSPC912795-5G
APOSPC912795-25G
2-(Difluoromethyl)-1-fluoro-4-nitrobenzene 95%
2-(Difluoromethyl)-1-fluoro-4-nitrobenzene
Formel:
C₇H₄F₃NO₂ |
MDL Number:
MFCD14698657 CAS-Nummer: 63878-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...