2-Aminoethyl methacrylate hydrochloride 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Aminoethyl 2-methylprop-2-enoate hydrochloride
OR61529-1G
OR61529-5G
OR61529-10G
APOSOR61529-1GEA
21.9
EUR
APOSOR61529-1G
APOSOR61529-5G
APOSOR61529-10G
2-Aminoethyl methacrylate hydrochloride 98%
2-Aminoethyl methacrylate hydrochloride
Formel:
C₆H₁₂ClNO₂ Schmelzpunkt: 102…110 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00078260 CAS-Nummer: 2420-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...