3-Bromanthranilsäure 90%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Amino-3-bromobenzoic acid
OR61456-25G
OR61456-100G
OR61456-500G
APOSOR61456-25GEA
43.5
EUR
APOSOR61456-25G
APOSOR61456-100G
APOSOR61456-500G
3-Bromanthranilsäure 90%
3-Bromanthranilsäure
Formel:
C₇H₆BrNO₂ |
MDL Number:
MFCD03618453 CAS-Nummer: 20776-51-6 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...