Lithium triisopropoxy(thiazol-2-yl)borate

Lieferant: Apollo Scientific
Achtung

OR72200-100MG OR72200-250MG OR72200-1G
APOSOR72200-100MGEA 65.4 EUR
APOSOR72200-100MG APOSOR72200-250MG APOSOR72200-1G
Lithium triisopropoxy(thiazol-2-yl)borate
Lithium triisopropoxy(thiazol-2-yl)borate
Formel: C₁₂H₂₃BLiNO₃S
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD16872058
CAS-Nummer: 1393823-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR