4-(2-Aminoethyl)cyclohexanol 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(2-Aminoethyl)cyclohexanol (cis- and trans- mixture)

OR938515-250MG OR938515-1G OR938515-5G
APOSOR938515-250MGEA 154 EUR
APOSOR938515-250MG APOSOR938515-1G APOSOR938515-5G
4-(2-Aminoethyl)cyclohexanol 95%
4-(2-Aminoethyl)cyclohexanol
Formel: C₈H₁₇NO
Siedepunkt: 256 °C
MDL Number: MFCD11520548
CAS-Nummer: 148356-06-3
UN-Nummer: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR