4-Cyclopropoxybenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR942399-500MG
OR942399-1G
APOSOR942399-500MGEA
683
EUR
APOSOR942399-500MG
APOSOR942399-1G
4-Cyclopropoxybenzoic acid 95%
4-Cyclopropoxybenzoic acid
Formel:
C₁₀H₁₀O₃ Schmelzpunkt: 153 - 155 °C |
MDL Number:
MFCD17000469 CAS-Nummer: 62577-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...