4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC305027-250MG
PC305027-1G
APOSPC305027-250MGEA
139
EUR
APOSPC305027-250MG
APOSPC305027-1G
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃BrF₄O₂ |
MDL Number:
MFCD31695851 CAS-Nummer: 2092502-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...