3-(Boc-amino)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

OR10346-1G OR10346-5G OR10346-25G
APOSOR10346-1GEA 62.5 EUR
APOSOR10346-1G APOSOR10346-5G APOSOR10346-25G
3-(Boc-amino)phenylboronsäure 97%
3-(Boc-amino)phenylboronsäure
Formel: C₁₁H₁₆BNO₄
Schmelzpunkt: 168 °C
MDL Number: MFCD03411945
CAS-Nummer: 380430-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR