2-Bromo-3-fluoro-5-iodopyridine 95%

Lieferant: Apollo Scientific
Achtung

PC49824-250MG PC49824-1G
APOSPC49824-250MGEA 261 EUR
APOSPC49824-250MG APOSPC49824-1G
2-Bromo-3-fluoro-5-iodopyridine 95%
2-Bromo-3-fluoro-5-iodopyridine
Formel: C₅H₂BrFIN
MDL Number: MFCD13185371
CAS-Nummer: 1214323-00-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR