2-Bromo-3-iodopyridin-4-amine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR62195-250MG
OR62195-1G
APOSOR62195-250MGEA
163
EUR
APOSOR62195-250MG
APOSOR62195-1G
2-Bromo-3-iodopyridin-4-amine 95%
2-Bromo-3-iodopyridin-4-amine
Formel:
C₅H₄BrIN₂ |
MDL Number:
MFCD28344964 CAS-Nummer: 1300750-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...