5-Bromo-2-methyl-4-nitropyridine-N-oxide 95%

Lieferant: Apollo Scientific
Achtung

OR929296-250MG OR929296-1G OR929296-5G
APOSOR929296-250MGEA 156 EUR
APOSOR929296-250MG APOSOR929296-1G APOSOR929296-5G
5-Bromo-2-methyl-4-nitropyridine-N-oxide 95%
5-Bromo-2-methyl-4-nitropyridine-N-oxide
Formel: C₆H₅BrN₂O₃
MDL Number: MFCD11977426
CAS-Nummer: 62516-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR