4-Bromo-2-(difluoromethyl)-1-iodobenzene 95%

Lieferant: Apollo Scientific
Gefahr

PC99170-250MG PC99170-1G PC99170-5G
APOSPC99170-250MGEA 190 EUR
APOSPC99170-250MG APOSPC99170-1G APOSPC99170-5G
4-Bromo-2-(difluoromethyl)-1-iodobenzene 95%
4-Bromo-2-(difluoromethyl)-1-iodobenzene
Formel: C₇H₄BrF₂I
MDL Number: MFCD18391112
CAS-Nummer: 1261819-63-9
UN-Nummer: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR