4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC99136-250MG
PC99136-1G
PC99136-5G
APOSPC99136-250MGEA
203
EUR
APOSPC99136-250MG
APOSPC99136-1G
APOSPC99136-5G
4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide
Formel:
C₈H₄Br₂F₄ |
MDL Number:
MFCD33404156 CAS-Nummer: 2090979-68-1 UN-Nummer: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...