4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC99133-250MG PC99133-1G PC99133-5G
APOSPC99133-250MGEA 174 EUR
APOSPC99133-250MG APOSPC99133-1G APOSPC99133-5G
4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
MDL Number: MFCD22052785
CAS-Nummer: 194804-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR