3-Bromo-5-chlorobenzyl bromide 96%

Lieferant: Apollo Scientific
Gefahr

OR400325-250MG OR400325-1G OR400325-5G
APOSOR400325-250MGEA 132 EUR
APOSOR400325-250MG APOSOR400325-1G APOSOR400325-5G
3-Bromo-5-chlorobenzyl bromide 96%
3-Bromo-5-chlorobenzyl bromide
Formel: C₇H₅Br₂Cl
MDL Number: MFCD09261035
CAS-Nummer: 762292-63-7
UN-Nummer: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR