3-Bromo-5-chloroisatoic anhydride 95%

Lieferant: Apollo Scientific
Achtung

OR400034-250MG OR400034-1G OR400034-5G
APOSOR400034-250MGEA 190 EUR
APOSOR400034-250MG APOSOR400034-1G APOSOR400034-5G
3-Bromo-5-chloroisatoic anhydride 95%
3-Bromo-5-chloroisatoic anhydride
Formel: C₈H₃BrClNO₃
MDL Number: MFCD11604568
CAS-Nummer: 122588-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR