5-Chlor-2-fluor-3-methylpyridin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Chloro-2-fluoro-3-picoline

PC9275-5G PC9275-25G PC9275-100G PC9275-500G
APOSPC9275-5GEA 34.8 EUR
APOSPC9275-5G APOSPC9275-25G APOSPC9275-100G APOSPC9275-500G
5-Chlor-2-fluor-3-methylpyridin 98%
5-Chlor-2-fluor-3-methylpyridin
Formel: C₆H₅ClFN
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03095094
CAS-Nummer: 375368-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR