Boc-statine

Lieferant: AAT BIOQUEST
Achtung

5100 5101
AATB5100EA 769 EUR
AATB5100 AATB5101
Boc-statine
Boc-statine

(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.

Formel: C₁₃H₂₅NO₅
MDL Number: MFCD00063180
CAS-Nummer: 58521-49-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d227ce2e7e469615ce301d8a46dbbcce516caa44&vid=a125260ea2eb10dafff9f31cd6a0079b7a572f91&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR