5(6)-TAMRA ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt
360
361
362
AATB360EA
152
EUR
AATB360
AATB361
AATB362
5(6)-TAMRA ≥95%
5(6)-TAMRA
TAMRA is one of the most popular fluorophores used in various bioconjugations.
Formel:
C₂₅H₂₂N₂O₅ Molekulargewicht: 430.45 g/mol |
MDL Number:
MFCD16661194 CAS-Nummer: 98181-63-6 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...