Calcein AM ≥95%, Ultrareine Qualität
Lieferant: AAT BIOQUEST
Achtung
Synonyme: Calcein-AM, Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein acetoxymethyl ester, Tetrakis(acetoxymethyl) 2,2',2'',2'''-(((3',6'-diacetoxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetate
22003 22004
AATB22003EA 230 EUR
AATB22003 AATB22004
Calcein AM ≥95%, Ultrareine Qualität
Calcein AM
Calcein AM readily passes through the cell membrane of viable cells because of its enhanced hydrophobicity as compared to calcein.
Formel: C₄₆H₄₆N₂O₂₃
Molekulargewicht: 994.86 g/mol
MDL Number: MFCD05861516
CAS-Nummer: 148504-34-1
ADR: 5 II